Learn More
Raybiotech Inc Human Osteoactivin (GPNMB) ELISA Kit
Supplier:  Raybiotech Inc ELHOSTEOACTIVIN1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 45 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This antibody pair detects human Osteoactivin. It show less than 20% cross-reactivity with recombinant mouse Osteoactivin.
Catalog No. 50-194-9609