Learn More
Abcam Human Osteocalcin ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB2702021X96TESTS
Encompass_Preferred
Human Osteocalcin ELISA Kit is a single-wash 90-min Simplestep used to quantify Human Osteocalcin with a sensitivity of 13.99 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair - Cited in over 10 citations
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1856332