Learn More
Biomatik Corporation Human Osteoprotegerin (OPG) CLIA Kit
Supplier:  Biomatik Corporation EKU08829
Encompass_Preferred
Human Osteoprotegerin (OPG) CLIA Kit for detecting OPG in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 41.2-30,000pg/ml. Sensitivity: 14.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: O00300. Gene ID: 4982. Target Synonyms: TNFRSF11B; TNFRSF11-B; OCIF; OPG; TR1; Tumor Necrosis Factor Receptor Superfamily Member 11b; Osteoclastogenesis Inhibitory Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7055