Learn More
American Research Products Inc Human Osteoprotegerin OPG ELI
Catalog No. 5000326574
Encompass
Human Osteoprotegerin OPG ELISA KIT from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26574 Supplier American Research Products Inc Supplier No. CSBE04692H
Human Osteoprotegerin OPG ELISA KIT from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates Method Sandwich