Learn More
Biomatik Corporation Human Ovarian Cancer Immuno Reactive Antigen Domain Containing Protein 2 (OCIAD2) ELISA Kit
Supplier:  Biomatik Corporation EKU06442
Encompass_Preferred
Human Ovarian Cancer Immuno Reactive Antigen Domain Containing Protein 2 (OCIAD2) ELISA Kit for detecting OCIAD2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.112ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q56VL3. Gene ID: 132299. Target Synonyms: OCIA Domain Containing 2; Ovarian carcinoma immunoreactive antigen-like protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7191