Learn More
Zyagen Labs Human ovary total RNA is a high pure intact total RNA isolated from freshly harvested normal healthy HUMAN OVARY tissues, treated with DNase and dissolved in water. RNA is ready for immediate use in any downstream application.
SDP
Catalog No. 501718104
Encompass_Preferred
Zyagen Total RNA is routinely extracted from freshly harvested normal healthy tissues of single donor using classical guanidine isothiocyanate–phenol:chloroform extraction method which allows the rapid isolation of total RNA including microRNAs. RNA is treated with RNase-free DNase to remove residual DNA, precisely quantified, and stored at -80oC. The integrity of each RNA sample as indicated by intact ribosomal RNA is verified by denatured agarose gel electrophoresis. The purity of RNA is assessed by NanoDrop (A260/A280: 1.9-2.1). Residual DNA contamination is tested by PCR. RNA is ideal for Northern blotting, ribonuclease protection assay, SI nuclease assay, RT-PCR/Q-PCR/RACE analysis, cDNA synthesis, RNA differential display, microRNA studies, and purification of mRNA for library construction. Total RNA sample is provided in RNase-free water at a concentration of 1 mg/ml and shipped on dry ice.
Catalog No. 50-171-8104 Supplier Zyagen Labs Supplier No. HR406
Zyagen Total RNA is routinely extracted from freshly harvested normal healthy tissues of single donor using classical guanidine isothiocyanate–phenol:chloroform extraction method which allows the rapid isolation of total RNA including microRNAs. RNA is treated with RNase-free DNase to remove residual DNA, precisely quantified, and stored at -80oC. The integrity of each RNA sample as indicated by intact ribosomal RNA is verified by denatured agarose gel electrophoresis. The purity of RNA is assessed by NanoDrop (A260/A280: 1.9-2.1). Residual DNA contamination is tested by PCR. RNA is ideal for Northern blotting, ribonuclease protection assay, SI nuclease assay, RT-PCR/Q-PCR/RACE analysis, cDNA synthesis, RNA differential display, microRNA studies, and purification of mRNA for library construction. Total RNA sample is provided in RNase-free water at a concentration of 1 mg/ml and shipped on dry ice.