Learn More
OriGene UNC13A Human Over-Expression Lysate, Positive Control for Western Blot, 100 µg
SDP
Supplier:  OriGene LY421638
Encompass_Preferred
The OriGene Technologies UNC13A Human Over-expression Lysate is a reliable positive control for protein-based assays like Western blots. Derived from HEK293T cells, this lysate features UNC13A protein tagged with C-Myc/DDK, ensuring consistent detection. Available in various formats, it supports diverse research needs, emphasizing dependable performance in detecting target proteins.
Catalog No. NC2822576