Learn More
Biomatik Corporation Human Oviductal Glycoprotein 1 (OVGP1) ELISA Kit
Supplier:  Biomatik Corporation EKU12451
Encompass_Preferred
Human Oviductal Glycoprotein 1 (OVGP1) ELISA Kit for detecting OVGP1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q12889. Target Synonyms: EGP; CHIT5; MUC9; OGP; Mucin 9; Oviductin; Oviduct-Specific Glycoprotein; Estrogen-dependent oviduct protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7347