Learn More
ACROBiosystems Human OX40 Ligand 1mg
Catalog No. 502534342
Encompass_Preferred
Human OX40 Ligand Protein His Tag (OXL-H5243) is expressed from human 293 cells (HEK293). It contains AA Gln 51 - Leu 183 (Accession # P23510-1).
Catalog No. 50-253-4342 Supplier ACROBiosystems Supplier No. OXLH52431MG
Human OX40 Ligand Protein His Tag (OXL-H5243) is expressed from human 293 cells (HEK293). It contains AA Gln 51 - Leu 183 (Accession # P23510-1).