Learn More
ACROBiosystems Biotinylated Human OX40 Ligand / TNFSF4 Protein, His,Avitag™, active trimer (MALS verified)
Catalog No. 502331659
Encompass_Preferred
Biotinylated Human OX40 Ligand, His,Avitag (OXL-H82Q5) is expressed from human 293 cells (HEK293). It contains AA Gln 51 - Leu 183 (Accession # P23510-1).
Catalog No. 50-233-1659 Supplier ACROBiosystems Supplier No. OXLH82Q5200UG
Biotinylated Human OX40 Ligand, His,Avitag (OXL-H82Q5) is expressed from human 293 cells (HEK293). It contains AA Gln 51 - Leu 183 (Accession # P23510-1).