Learn More
ACROBiosystems HEK293/Human OX40 Ligand Stable Cell Line, Adherent, 2/Pk.
Catalog No. 502331302
Encompass_Preferred
This HEK293 stable cell line is engineered to express the human OX40 Ligand (TNFSF4). It is designed to serve as a reliable tool for studying OX40 Ligand interactions and supporting cell-based bioassays.
  • Adherent growth properties
  • Validated for binding assays via FACS and cell-based ELISA
  • Maintained in DMEM medium supplemented with 10% FBS
  • Tested negative for mycoplasma and sterility
  • Selected using puromycin at 10 μg/mL
  • Supplied at a concentration of at least 5x106 cells per vial in 10% DMSO and 90% FBS
Catalog No. 50-233-1302 Supplier ACROBiosystems Supplier No. CHEKATP0542VIALS
This HEK293 stable cell line is engineered to express the human OX40 Ligand (TNFSF4). It is designed to serve as a reliable tool for studying OX40 Ligand interactions and supporting cell-based bioassays.
  • Adherent growth properties
  • Validated for binding assays via FACS and cell-based ELISA
  • Maintained in DMEM medium supplemented with 10% FBS
  • Tested negative for mycoplasma and sterility
  • Selected using puromycin at 10 μg/mL
  • Supplied at a concentration of at least 5x106 cells per vial in 10% DMSO and 90% FBS