Learn More
ACROBiosystems Human OX40 Ligand 50ug
Supplier:  ACROBiosystems OXLH524350UG
Encompass_Preferred
Human OX40 Ligand Protein His Tag (OXL-H5243) is expressed from human 293 cells (HEK293). It contains AA Gln 51 - Leu 183 (Accession # P23510-1).
Catalog No. 50-253-4341