Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human OX40 Ligand (soluble) Recombinant Protein, PeproTech®

Catalog No. 3102810UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
310-28-10UG 10 μg
31028100UG 100 μg
310-28-1MG 1 mg
31028250UG 250 μg
310-28-2UG 2 μg
31028500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 310-28-10UG Supplier Gibco™ Supplier No. 3102810UG
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant OX40L is a glycosylated 133 amino acid protein corresponding to the extracellular TNF homologous domain of the full length transmembrane protein. It migrates with an apparent molecular mass of 15.5-25.0 kDa on SDS-PAGE. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The tumor necrosis factor superfamily member TNFSF4 is a type II membrane bound, non-covalently linked homotrimeric protein. It is expressed on antigen presenting cells, such as dendritic cells and activated B-cells, and also on other cells such as vascular endothelial cells, mast cells, and natural killer cells. TNFSF4 signals specifically through the TNFRSF4 receptor, is expressed predominantly on CD4+T cells but also on certain activated CD8+T cells. TNFRSF4/TNFSF4 functions as a costimulatory signal, which is required for a productive interaction between antigen presenting cells and their target T-cells. It enhances cell proliferation and survival, and increases expression of RANTES, IL-2, IL-3, and IFNgamma. TNFRSF4/TNFSF4 signaling plays an important role in immunotolerance.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P23510
For Use With (Application) Control, ELISA Standard, Binding Assay, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 7292
Molecular Weight (g/mol) 15.4 kDa
Name Human OX40 Ligand (soluble)
Purification Method Purified
Quantity 10 μg
Source Insect cells
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias Ath1; Ath-1; atherosclerosis 1; CD134 ligand; CD134L; CD252; glycoprotein Gp34; GP34; OX40 antigen ligand; OX40 ligand; OX40L; OX-40L; OX4OL; TAX transcriptionally-activated glycoprotein 1; tax-transcriptionally activated glycoprotein 1 (34kD); tax-transcriptionally activated glycoprotein 1 ligand; TNFSF4; TNLG2B; tumor necrosis factor (ligand) superfamily member 4; tumor necrosis factor (ligand) superfamily, member 4; tumor necrosis factor ligand 2B; tumor necrosis factor ligand superfamily member 4; tumor necrosis factor superfamily member 4; TXGP1; Txgp1l
Common Name OX40L (CD252)
Gene Symbol Tnfsf4
Biological Activity Determined by its ability to bind hOX40 in a functional ELISA.
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence QVSHRYPRIQ SIKVQFTEYK KEKGFILTSQ KEDEIMKVQN NSVIINCDGF YLISLKGYFS QEVNISLHYQ KDEEPLFQLK KVRSVNSLMV ASLTYKDKVY LNVTTDNTSL DDFHVNGGEL ILIHQNPGEF CVL
Content And Storage -20°C
Activity Determined by its ability to bind hOX40 in a functional ELISA.
Expression System Insect cells
Form Lyophilized
Purity or Quality Grade ≥ 97% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.