Learn More
Raybiotech Inc Human OX40/TNFRSF4 ELISA Kit
Supplier:  Raybiotech Inc ELHOX401
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 3.2 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human OX40.
Catalog No. 50-194-9612