Learn More
BIOHIPPO Human OxHDL Microsample
SDP
Catalog No. 5000315219
Encompass
Human OxHDL Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of OxHDL in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 1 57-100 ng/mL with a sensitivity of 0 6 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human OxHDL Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human OxHDL Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human OxHDL biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15219 Supplier BIOHIPPO Supplier No. BHE15210577
Human OxHDL Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of OxHDL in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 1 57-100 ng/mL with a sensitivity of 0 6 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human OxHDL Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human OxHDL Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human OxHDL biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color