Learn More
Biomatik Corporation Human Oxysterol Binding Protein Like Protein 8 (OSBPL8) ELISA Kit
Supplier:  Biomatik Corporation EKU06453
Encompass_Preferred
Human Oxysterol Binding Protein Like Protein 8 (OSBPL8) ELISA Kit for detecting OSBPL8 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.138ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9BZF1. Gene ID: 114882. Target Synonyms: ORP8; OSBP10; Oxysterol-Binding Protein-Related Protein 8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7184