Learn More
BIOHIPPO Human P-Cadherin Microsample
SDP
Catalog No. 5000313817
Encompass
Human P-Cadherin Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of P-Cadherin in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 62 50-4000 pg/mL with a sensitivity of 36 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human P-Cadherin Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human P-Cadherin Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human P-Cadherin biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13817 Supplier BIOHIPPO Supplier No. BHE15210514
Human P-Cadherin Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of P-Cadherin in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 62 50-4000 pg/mL with a sensitivity of 36 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human P-Cadherin Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human P-Cadherin Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human P-Cadherin biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color