Learn More
Raybiotech Inc Human P-Selectin ELISA Kit
Supplier:  Raybiotech Inc ELHPSELECTIN1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 20 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA kit shows no significant cross-reactivity with any of the following cytokines tested: human Angiogenin, BDNF, BLC, CNTF, EGF, ENA-78, EOT, FGF-4, FGF-6, FGF-7, IL-1 alpha, IL-1 beta, IL-4, IL-5, IL-11, IL-12 p70, IL-12 p40, IL-13, I-309, IP-10, M-CSF, IFN-gamma, MCP- 1, MCP-2, MCP-3, MDC, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, SCF, TARC, TIMP-1, TIMP-2, TPO, VEGF.
Catalog No. 50-194-9667