Learn More
Biomatik Corporation Human p21 Protein Activated Kinase 4 (PAK4) CLIA Kit
Supplier:  Biomatik Corporation EKU10038
Encompass_Preferred
Human p21 Protein Activated Kinase 4 (PAK4) CLIA Kit for detecting PAK4 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 54.87-40,000pg/ml. Sensitivity: 19.28pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: O96013. Gene ID: 10298. Target Synonyms: Serine/threonine-protein kinase PAK 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7662