Learn More
Bioworld HUMAN P21-WAF1 (RECOMBINANT)
Supplier:  Bioworld 220606141
Encompass_Preferred
Cyclin-dependent kinase inhibitor 1(CDKN1A/P21/WAF1) Source: E.Coli Background: p21/WAF1/Cip1/Sdi1/Pic1 is a tumor suppressor protein. Expression of p21WAF1 is induced by wild type, but not mutant, p53 suppressor protein. The p21/WAF...
Catalog No. 50-253-714