Learn More
BIOHIPPO Human P2RY12 Microsample
SDP
Catalog No. 5000315038
Encompass
Human P2RY12 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of P2RY12 in tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 057 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human P2RY12 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human P2RY12 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human P2RY12 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15038 Supplier BIOHIPPO Supplier No. BHE15211521
Human P2RY12 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of P2RY12 in tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 057 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human P2RY12 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human P2RY12 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human P2RY12 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color