Learn More
Bioworld HUMAN P38 ALPHA/SAPK2 ALPHA
Supplier:  Bioworld 220604341
Encompass_Preferred
HUMAN P38 ALPHA/SAPK2 ALPHA (RECOMBINANT): Recombinant Human JNK2/SAPK1 Source: E.Coli Description : Recombinant p38/SAPK2 is a non-glycosylated polypeptide produced by phosphorylation of the purified p38 alpha with MKK6 having a mol...
Catalog No. 50-253-531