Learn More
Sino Biological Human P3H2 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 81412R08H50
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human LEPREL1 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-237-4107