Learn More
Raybiotech Inc Human P4HB ELISA Kit
Supplier:  Raybiotech Inc ELHP4HB1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.22 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human P4HB.
Catalog No. 50-194-9614