Learn More
Sino Biological Recombinant Anti-Protein disulfide-isomerase/PDI Antibody (FITC), Rabbit Monoclonal
Supplier:  Sino Biological 10827R016F100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human P4HB (rh P4HB; Catalog#10827-H08H; NP_000909.2; Met 1-Lys 505) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-241-1617