Learn More
Sino Biological Anti-Protein disulfide-isomerase/PDI Antibody (HRP), Mouse Monoclonal
Supplier:  Sino Biological 10827MM08H100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human P4HB / ERBA2L (rh P4HB; Catalog#10827-H08H; NP_000909.2; Met 1-Lys 505). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-241-0794