Learn More
Sino Biological Anti-Protein disulfide-isomerase/PDI Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10827MM03100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human P4HB (rh P4HB; Catalog#10827-H08H; NP_000909.2; Met 1-Lys 505). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-0393