Learn More
Sino Biological Human P4HB PAb 100ul (Sino Biological)
Supplier:  Sino Biological 10827RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human P4HB / ERBA2L (rh P4HB; Catalog#10827-H08H; NP_000909.2; Met 1-Lys 505). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-237-2138