Learn More
Abcam Human p53 ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1715711X96TESTS
Encompass_Preferred
Human p53 ELISA Kit is a single-wash 90-min Simplestep used to quantify Human p53 with a sensitivity of 65 pg/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Cited in over 10 citations
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2007777