Learn More
Abcam Human p53 ELISA Kit (pSer15), 1 x 96 Tests
Supplier:  Abcam AB1560271X96TESTS
Encompass_Preferred
Human p53 ELISA Kit (pSer15) is a Sandwich (quantitative) ELISA kit for the measurement of Human p53 (pSer15) in Human in Tissue Lysate, Cell culture extracts, Suspension cells, Tissue Homogenate, Adherent cells, Cell Lysate samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2824237