Learn More
Biomatik Corporation Human P62/SQSTM1 (Sequestosome-1) ELISA Kit
Supplier:  Biomatik Corporation EKF58867
Encompass_Preferred
Human P62/SQSTM1 (Sequestosome-1) ELISA Kit for detecting P62; SQSTM1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q13501. Target Synonyms: A170; EBIAP; ORCA; OSIL; p60; P62; P62;SQSTM1; p62B; PDB3; sequestosome 1; SQSTM1; Ubiquitin binding protein p62; ZIP3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5872