Learn More
BIOHIPPO Human PA2G4 Microsample
SDP
Catalog No. 5000315440
Encompass
Human PA2G4 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PA2G4 in tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 114 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human PA2G4 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human PA2G4 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human PA2G4 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15440 Supplier BIOHIPPO Supplier No. BHE15211187
Human PA2G4 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PA2G4 in tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 114 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human PA2G4 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human PA2G4 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human PA2G4 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color