Learn More
BIOHIPPO Human PABPC1L Microsample
SDP
Catalog No. 5000315164
Encompass
Human PABPC1L Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PABPC1L in Tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 118 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human PABPC1L Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human PABPC1L Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human PABPC1L biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15164 Supplier BIOHIPPO Supplier No. BHE15210975
Human PABPC1L Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PABPC1L in Tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 118 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human PABPC1L Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human PABPC1L Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human PABPC1L biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color