Learn More
Sino Biological Human PADI4 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 11072RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human PAD4 / PADI4 (rh PAD4 / PADI4; Catalog#11072-H07B; ; Met1-Pro663). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-237-3097