Learn More
Vector Laboratories Human PAF / Platelet Activating Factor Protein (Recombinant) - LS-G126, 20ug
SDP
Catalog No. NC3881127
Encompass
PAF / Platelet Activating Factor Protein LS-G126 is a Recombinant Human PAF / Platelet Activating Factor produced in HEK 293 Cells. It is biologically active and is low in endotoxin; Less than 0.1 ng/µg of protein (less than 1EU/µg). For Research Use Only
Catalog No. NC3881127 Supplier Vector Laboratories Supplier No. LSG12620
PAF / Platelet Activating Factor Protein LS-G126 is a Recombinant Human PAF / Platelet Activating Factor produced in HEK 293 Cells. It is biologically active and is low in endotoxin; Less than 0.1 ng/µg of protein (less than 1EU/µg). For Research Use Only