Learn More
Biomatik Corporation Human Paired Box Gene 6 (PAX6) ELISA Kit
Supplier:  Biomatik Corporation EKU12354
Encompass_Preferred
Human Paired Box Gene 6 (PAX6) ELISA Kit for detecting PAX6 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P26367. Target Synonyms: AN; AN2; MGDA; WAGR; Oculorhombin; Aniridia,Keratitis; Aniridia type II protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5527