Learn More
BIOHIPPO Human PAK1 Microsample
SDP
Catalog No. 5000313967
Encompass
Human PAK1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PAK1 in Serum plasma and other biological fluids It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 6 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human PAK1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human PAK1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human PAK1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13967 Supplier BIOHIPPO Supplier No. BHE15210623
Human PAK1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PAK1 in Serum plasma and other biological fluids It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 6 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human PAK1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human PAK1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human PAK1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color