Learn More
Biomatik Corporation Human Pancreas Specific Transcription Factor 1a (PTF1a) ELISA Kit
Supplier:  Biomatik Corporation EKU09136
Encompass_Preferred
Human Pancreas Specific Transcription Factor 1a (PTF1a) ELISA Kit for detecting PTF1a in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q7RTS3. Gene ID: 256297. Target Synonyms: PTF1-A; PTF1-p48; Class A basic helix-loop-helix protein 29; bHLH transcription factor p48; p48 DNA-binding subunit of transcription factor PTF1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6854