Learn More
Biomatik Corporation Human Pancreatic Lipase (PL) ELISA Kit
Supplier:  Biomatik Corporation EKU08336
Encompass_Preferred
Human Pancreatic Lipase (PL) ELISA Kit for detecting PL in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.269ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P16233. Gene ID: 5406. Target Synonyms: PNLIP; PTL; Pancreatic Lipase; Pancreatic triacylglycerol lipase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5822