Learn More
Cell Systems Corporation Primary Human Pancreatic Microvascular Endothelial Cells (ACBRI 134 V)
SDP
Catalog No. NC1686614
Encompass_Preferred
Primary Human Pancreatic Microvascular Endothelial Cells (ACBRI 134 V) were initiated from normal human pancreatic tissue. Isolated from normal human pancreatic tissue and, unlike cells from other providers, without positive control antibody labels. Each vial contains ~1 x 10^6 cells with 85-92% viability under ideal conditions. They are available at Passage 3 (<12 cumulative population doublings) and will initiate a Passage 4 cell culture in a 75 cm2 flask. Each vial of cells is shipped with Bac-Off (antibiotic) and CultureBoost (animal derived growth factors) at no additional cost. You can enhance consistency in your research program with reserved lots. Need vials in bulk? Contact your Fisher Scientific Sales representative for tailored quotes. Tight Deadline? Our cells are available for next day delivery after the purchase order is received. Accelerate Your Research! Try our Attachment Factor (4Z0-201, 4Z0-210) – innovative 5 seconds plating solution for all surfaces.
Catalog No. NC1686614 Supplier Cell Systems Corporation Supplier No. ACBRI 134 V
Add to Cart
Add to Cart
Primary Human Pancreatic Microvascular Endothelial Cells (ACBRI 134 V) were initiated from normal human pancreatic tissue. Isolated from normal human pancreatic tissue and, unlike cells from other providers, without positive control antibody labels. Each vial contains ~1 x 10^6 cells with 85-92% viability under ideal conditions. They are available at Passage 3 (<12 cumulative population doublings) and will initiate a Passage 4 cell culture in a 75 cm2 flask. Each vial of cells is shipped with Bac-Off (antibiotic) and CultureBoost (animal derived growth factors) at no additional cost. You can enhance consistency in your research program with reserved lots. Need vials in bulk? Contact your Fisher Scientific Sales representative for tailored quotes. Tight Deadline? Our cells are available for next day delivery after the purchase order is received. Accelerate Your Research! Try our Attachment Factor (4Z0-201, 4Z0-210) – innovative 5 seconds plating solution for all surfaces.