Learn More
Cusabio Technology LLC Human Pantetheinase VNN1 ELISA
Catalog No. 5000049408
Encompass_Preferred
This kit detects VNN1 (Human Pantetheinase) in serum plasma and tissue homogenates with a dynamic range of 78 pg/mL to 5000 pg/mL offering high sensitivity down to 19 5 pg/mL Designed for quantitative analysis it employs a sandwich ELISA technique that delivers results within 1-5 hours using 50-100 L of sample per test The assay measures absorbance at 450 nm to accurately quantify VNN1 concentrations making it suitable for both research and diagnostic applications requiring precise detection of this enzyme in various biological matrices Its optimized protocol ensures reliable performance across the specified detection range while maintaining consistency in experimental outcomes
Catalog No. 50-000-49408 Supplier Cusabio Technology LLC Supplier No. CSBEL025883HU
This kit detects VNN1 (Human Pantetheinase) in serum plasma and tissue homogenates with a dynamic range of 78 pg/mL to 5000 pg/mL offering high sensitivity down to 19 5 pg/mL Designed for quantitative analysis it employs a sandwich ELISA technique that delivers results within 1-5 hours using 50-100 L of sample per test The assay measures absorbance at 450 nm to accurately quantify VNN1 concentrations making it suitable for both research and diagnostic applications requiring precise detection of this enzyme in various biological matrices Its optimized protocol ensures reliable performance across the specified detection range while maintaining consistency in experimental outcomes