Learn More
ATCC HPV type 6b; purified plasmid DNA
Supplier:  ATCC 45150D
Encompass
This is a clone of human papilloma virus type 6b that has been linearized at the unique BamHI site (nt 4722) in the coding region for L2 and cloned into the BamHI site of the pUC19 vector.
Catalog No. 50-237-9658