Learn More
Raybiotech Inc Human PAR1 ELISA Kit
Supplier:  Raybiotech Inc ELHPAR11
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 3 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA kit shows no cross-reactivity with the following cytokines tested: human Coagulation Factor X, CHMP2B, Contactin-3, Cortactin, CrkL, Cyr61/CCN1, DAPP1, Dynactin Subunit 1/DCTN1, DFF45/ICAD, DRAK1, GRAP2, GRK5, Hydroxyacid Oxidase- 1/HAO-1, LRIG1, MMP-12, NCK1, Nectin-2/CD112, Nesfatin- 1/Nucleobindin-2, Neurogranin, Nrf2, NUDT5, Nucleoporin NUP85, Pancreatic Polypeptide/PP, Peroxiredoxin 2, PSMA1, PU.1/Spi-1, RalA, RCOR1/CoREST, Serpin B8, SH2D1A, SHP-1, Siglec-6/CD327, SorCS3, THAP11, ULBP- 4/RAET1E, UNC5H3, VAMP-1, VAMP-2, PBEF/Visfatin.
Catalog No. 50-194-9619