Learn More
ATCC Human parainfluenza virus 1; (HPIV-1); Strain: C35
Supplier:  ATCC VR94
Encompass
Human parainfluenza virus 1 strain C35 is propagated in LLC-MK2 Derivative cells (ATCC CCL-7.1). This strain was isolated in 1957 from the throat swab of a 3-year-old human male with acute laryngitis in Washinton, DC. This product is whole-genome sequenced and has applications in respiratory disease research.
Catalog No. 50-238-2394