Learn More
ATCC Human parainfluenza; virus 1 RNA; Strain: C35
Supplier:  ATCC VR94D
Encompass
Genomic RNA isolated from LLC-MK2 cells (ATCC CCL-7.1) infected with Human parainfluenza virus 1 strain C35 (ATCC VR-94).This product was prepared using methods known to inactivate viruses. It is suitable for use in RT-PCR or other RNA-based procedures. Source organism and host cells are also available through the ATCC Catalog.
Catalog No. 50-238-3174
 Restricted Item