Learn More
ATCC Human parainfluenza; virus 3, (HPIV-3); Strain: C 243
Catalog No. 502382991
Encompass
Human parainfluenza virus 3 strain C 243 is propagated in LLC-MK2 Derivative cells (ATCC CCL-7.1). This strain was isolated in 1957 from a one-year-old human female with pneumonia in Washington, DC, and was deposited by the Centers for Disease Control and Prevention. This product has applications in infectious and respiratory disease research.
Catalog No. 50-238-2991 Supplier ATCC Supplier No. VR93
Human parainfluenza virus 3 strain C 243 is propagated in LLC-MK2 Derivative cells (ATCC CCL-7.1). This strain was isolated in 1957 from a one-year-old human female with pneumonia in Washington, DC, and was deposited by the Centers for Disease Control and Prevention. This product has applications in infectious and respiratory disease research.