Learn More
ATCC Human parainfluenza; virus 3 RNA; Strain: C 243
Catalog No. 502384213
Encompass
Genomic RNA isolated from a preparation of cell lysate and supernatant from LLC-MK2 Derivative cells (ATCC CCL-7.1) infected with Human parainfluenza virus 3 strain C 243 (ATCC VR-93). This product was prepared using methods known to inactivate viruses. It is suitable for use in RT-PCR or other RNA-based procedures. The source organism and host cells are also available through the ATCC catalog.
Catalog No. 50-238-4213 Supplier ATCC Supplier No. VR93D
Genomic RNA isolated from a preparation of cell lysate and supernatant from LLC-MK2 Derivative cells (ATCC CCL-7.1) infected with Human parainfluenza virus 3 strain C 243 (ATCC VR-93). This product was prepared using methods known to inactivate viruses. It is suitable for use in RT-PCR or other RNA-based procedures. The source organism and host cells are also available through the ATCC catalog.