Learn More
ATCC Human parainfluenza; virus 4b; Strain: CH 19503
Catalog No. 502384069
Encompass
Human parainfluenza virus 4b strain CH 19503 was isolated from a child with a respiratory tract infection in Washington, DC, and was deposited by the Centers for Disease Control and Prevention. This animal virus can be propagated in NCI-H292 [H292] cells (ATCC CRL-1848) and has applications in infectious disease and respiratory disease research.
Catalog No. 50-238-4069 Supplier ATCC Supplier No. 502384069
Human parainfluenza virus 4b strain CH 19503 was isolated from a child with a respiratory tract infection in Washington, DC, and was deposited by the Centers for Disease Control and Prevention. This animal virus can be propagated in NCI-H292 [H292] cells (ATCC CRL-1848) and has applications in infectious disease and respiratory disease research.