Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Human Paraoxonase 1 (PON) Standards
SDP

Catalog No. 22157379 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
2 x 0.3 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-157-379 2 x 0.3 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-157-379 Supplier ZeptoMetrix Supplier No. 0801384
Add to Cart
Add to Cart

Used as controls for arylesterase and paraoxonase applications, including kinetic activity assays and phenotype/genotype assays

  • Each package contains two vials: 300μL of both PON phenotype QQ and phenotype RR
  • Suspended in buffer containing 25mM Tris-HCl, 0.17mM NaCl, 20% glycerol, 0.1% Tergitol NP-10, 1.0mM CaCl2, at pH 8.0

Specifications

Content And Storage -70°C
Form Solution
Media Type Vial
Product Type Human Paraoxonase 1 Standard
For Use With (Application) Used for control
Quantity 2 x 0.3 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.