Learn More
Biomatik Corporation Human Parathyroid Hormone Receptor 2 (PTHR2) ELISA Kit
Supplier:  Biomatik Corporation EKU06487
Encompass_Preferred
Human Parathyroid Hormone Receptor 2 (PTHR2) ELISA Kit for detecting PTHR2 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.118ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P49190. Gene ID: 5746. Target Synonyms: PTH2; PTH2R; Parathyroid Hormone 2 Receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7581